1-Eicosanol, 98%
1-Eicosanol, 98%
1-Eicosanol, 98%
Thermo Scientific Chemicals

1-Eicosanol, 98%

CAS: 629-96-9 | C20H42O | 298.56 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 215460250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS629-96-9
IUPAC Nameicosan-1-ol
Molecular FormulaC20H42O
InChI KeyBTFJIXJJCSYFAL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCO
さらに表示
Appearance (Color)White
Melting point62°C to 66°C
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder or pellets and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only