Tripalmitin, 99%
Tripalmitin, 99%
Tripalmitin, 99%
Thermo Scientific Chemicals

Tripalmitin, 99%

CAS: 555-44-2 | C51H98O6 | 807.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 215670250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS555-44-2
IUPAC Name1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular FormulaC51H98O6
InChI KeyPVNIQBQSYATKKL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
HPLC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only