Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals

Phenylmethanesulfonyl fluoride, 99%

CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
数量:
1 g
5 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 215740100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS329-98-6
IUPAC Namephenylmethanesulfonyl fluoride
Molecular FormulaC7H7FO2S
InChI KeyYBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILESFS(=O)(=O)CC1=CC=CC=C1
さらに表示
Appearance (Form)Needles and/or agglomerates
Melting point90°C to 94°C
Appearance (Color)White
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only