BS(PEG)9 - FAQs
View additional product information for BS(PEG)9 - FAQs (21582)
2 product FAQs found
Can you provide the shelf-life for BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate)?
BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Do you recommend crosslinking reagents for viral capsids (diameter 30 nm)?
Amine-to-amine crosslinkers are a good choice for your application since amino groups are usually abundant on the surface of proteins. Our most popular amine-reactive homobifunctional crosslinker that is water-soluble is BS3 (Cat. No. A39266). This has a moderate-length (11.4 A) linker that is good for linking adjacent proteins. If you need the ability to cleave the linker with sulfhydryl reducing agents I would suggest DTSSP (Cat. No. 21578). If you think you might need an extra-long crosslinker, then BS(PEG)5 (Cat. No. 21581) or BS(PEG)9 (Cat. No. 21582) are alternatives.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.