Ethanesulfonyl chloride, 98+%
Ethanesulfonyl chloride, 98+%
Ethanesulfonyl chloride, 98+%
Thermo Scientific Chemicals

Ethanesulfonyl chloride, 98+%

CAS: 594-44-5 | C2H5ClO2S | 128.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 215820500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS594-44-5
IUPAC Nameethanesulfonyl chloride
Molecular FormulaC2H5ClO2S
InChI KeyFRYHCSODNHYDPU-UHFFFAOYSA-N
SMILESCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Form)Liquid
AppearanceMay darken on storage
Appearance (Color)Clear colorless to yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only