Butyl acrylate, 99+%, stabilized
Butyl acrylate, 99+%, stabilized
Butyl acrylate, 99+%, stabilized
Thermo Scientific Chemicals

Butyl acrylate, 99+%, stabilized

CAS: 141-32-2 | C7H12O2 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 215860010
Price (USD)
75.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
75.00
Each
Chemical Identifiers
CAS141-32-2
IUPAC Namebutyl prop-2-enoate
Molecular FormulaC7H12O2
InChI KeyCQEYYJKEWSMYFG-UHFFFAOYSA-N
SMILESCCCCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.0 %
Acidity=<0.01 % (as Acrylic acid)
Appearance (Form)Clear liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only