Product Image
Thermo Scientific™
Nalgene™ Polypropylene, Replacement Coupling Inserts, elbow coupling, 1/4 inch
For Use With: Nalgene Quick Filling/Venting Closures
Have Questions?
Catalog NumberQuantity
2159-0011Case of 6
Catalog number 2159-0011
Price (USD)
149.00
Case of 6
Quantity:
Case of 6
Price (USD)
149.00
Case of 6
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies
Specifications
Description90° Elbow coupling insert
Diameter (English) Inner Tubing1/4 in.
Diameter (Metric) Inner Tubing6.35 mm
For Use With (Equipment)Nalgene Quick Filling/Venting Closures
No. per Case6/Cs.
No. per Pack6 Pk.
QuantityCase of 6
MaterialPolypropylene
Product TypeInsert
Unit SizeCase of 6