DPBS, powder, no calcium, no magnesium, 50 L - FAQs
DPBS, powder, no calcium, no magnesium, 50 L - FAQs
View additional product information for DPBS, powder, no calcium, no magnesium - FAQs (21600069, 21600010, 21600044)
2 product FAQs found
Can I get a specific value for the conductivity for your DPBS products?
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Are your DPBS products DNase/RNase-free? Can I use them for RNA work?
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.