ART REACH Barrier Tips, Extended Length
Molecular Bio Products Inc.
ART REACH Barrier Tips, Extended Length
Precise liquid handling with contamination protection, featuring extended reach and secure racks for efficiency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
2160P-HR200 μL
214010 μL
Catalog number 2160P-HR
Price (USD)
672.00
Online exclusive
960.00
Save 288.00
Case of 3840
Volume (Metric):
200 μL
Price (USD)
672.00
Online exclusive
960.00
Save 288.00
Case of 3840
Precise liquid handling with contamination protection, featuring extended reach and secure racks for efficiency.
  • ART Reach Barrier Tips ensure accurate pipetting
  • Barrier filter prevents cross-contamination
  • Extended length design improves access to deep vessels
  • Hinged rack allows easy, one-handed operation
  • Sterile format suitable for sensitive applications
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART Barrier, Filtered, Sterile, Hinged Rack
Length (English)3.6 in.
Length (Metric)9.09 cm
ModelART Barrier
Product TypePipette Tips
Quantity960 Tips
AutoclavableNot Autoclavable
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)200 μL
Unit SizeCase of 3840