Tetraethylammonium fluoride hydrate, 98%
Tetraethylammonium fluoride hydrate, 98%
Tetraethylammonium fluoride hydrate, 98%
Thermo Scientific Chemicals

Tetraethylammonium fluoride hydrate, 98%

CAS: 98330-04-2 | C8H20FN | 149.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 216660250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98330-04-2
IUPAC Nametetraethylazanium fluoride
Molecular FormulaC8H20FN
InChI KeyQSUJAUYJBJRLKV-UHFFFAOYSA-M
SMILES[F-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystals or adhering crystals or crystalline solid
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Water=<28 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only