Iron(III) chloride hexahydrate, 99+%, for analysis
Iron(III) chloride hexahydrate, 99+%, for analysis
Iron(III) chloride hexahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 99+%, for analysis

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 217090025
Price (USD)
553.65
Special offer
Online exclusive
Ends: 28-Dec-2026
651.00
Save 97.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
553.65
Special offer
Online exclusive
Ends: 28-Dec-2026
651.00
Save 97.35
Each
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder and/or lumps
Phosphorous compounds (as PO4)=<100 ppm
Zinc (Zn)=<30 ppm
Ferrous iron (Fe 2+)=<20 ppm
Titration Iodometric99.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid for analysis
RUO – Research Use Only