n-Pentane, 99+%, extra pure, anhydrous
n-Pentane, 99+%, extra pure, anhydrous
n-Pentane, 99+%, extra pure, anhydrous
Thermo Scientific Chemicals

n-Pentane, 99+%, extra pure, anhydrous

CAS: 109-66-0 | C5H12 | 72.15 g/mol
Quantity:
1 L
2.5 L
4 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 217240251
Price (JPY)
-
Quantity:
25 L
Request bulk or custom format
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3570 to 1.3590 (20°C, 589 nm)
Infrared spectrumConforms
Residue after evaporation=<0.001 %
Appearance (Form)Clear liquid
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only