Chloroform-d, for NMR, 100.0% atom D, packaged in 0.50mL ampoules
Chloroform-d, for NMR, 100.0% atom D, packaged in 0.50mL ampoules
Chloroform-d, for NMR, 100.0% atom D, packaged in 0.50mL ampoules
Thermo Scientific Chemicals

Chloroform-d, for NMR, 100.0% atom D, packaged in 0.50mL ampoules

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
製品番号(カタログ番号)数量
2173200505 mL
製品番号(カタログ番号) 217320050
価格(JPY)
-
数量:
5 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
さらに表示
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Water=<0.01 % (Coulometric)
D-Enrichment>=99.95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only