Potassium hydrogen carbonate, 99.5+%, for analysis
Potassium hydrogen carbonate, 99.5+%, for analysis
Potassium hydrogen carbonate, 99.5+%, for analysis
Thermo Scientific Chemicals

Potassium hydrogen carbonate, 99.5+%, for analysis

CAS: 298-14-6 | CHKO3 | 100.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 217590010
Price (USD)
113.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with HCl99.5 to 100.5 %
Heavy metals (as Pb)=<5 ppm
Nitrogen compounds (as N)=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only