Lithium fluoride, 97%, for analysis
Lithium fluoride, 97%, for analysis
Lithium fluoride, 97%, for analysis
Thermo Scientific Chemicals

Lithium fluoride, 97%, for analysis

CAS: 7789-24-4 | FLi | 25.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 218270025
Price (USD)
791.65
Special offer
Online exclusive
Ends: 28-Dec-2026
931.00
Save 139.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
791.65
Special offer
Online exclusive
Ends: 28-Dec-2026
931.00
Save 139.35
Each
Chemical Identifiers
CAS7789-24-4
IUPAC Namelithium(1+) fluoride
Molecular FormulaFLi
InChI KeyPQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES[Li+].[F-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=96.0 %
Chloride (Cl)=<0.07 %
Phosphate (PO4)=<0.04 %
Iron (Fe)=<0.03 %
Sodium (Na)=<0.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only