Cesium sulfate, 99+%, pure
Cesium sulfate, 99+%, pure
Cesium sulfate, 99+%, pure
Thermo Scientific Chemicals

Cesium sulfate, 99+%, pure

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 218702500
Price (JPY)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10294-54-9
IUPAC Namedicaesium(1+) sulfate
Molecular FormulaCs2O4S
InChI KeyFLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Assay>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only