Search
View additional product information for RPMI 1640 Medium - FAQs (21875042)
2 product FAQs found
While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.