Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Thermo Scientific Chemicals

Drierite™, without indicator, 8 mesh

CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 219050020
Price (USD)
162.00
Each
Quantity:
2 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
162.00
Each
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Composition>=99.5 % (CaSO4) Typical
Appearance (Color)White to off-white
Appearance (Form)Irregular granules
Particle size8 mesh
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only