Drierite™, without indicator, 10-20 mesh
Drierite™, without indicator, 10-20 mesh
Drierite™, without indicator, 10-20 mesh
Thermo Scientific Chemicals

Drierite™, without indicator, 10-20 mesh

CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 219060020
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Quantity:
2 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Water=<0.3 %
Particle size10 to 20 mesh
Appearance (Form)Irregular granules
Composition>=99.5 % (CaSO4) Typical
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only