Calcium hydroxide, 98%, extra pure
Calcium hydroxide, 98%, extra pure
Calcium hydroxide, 98%, extra pure
Thermo Scientific Chemicals

Calcium hydroxide, 98%, extra pure

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 219180010
Price (USD)
94.65
Special offer
Online exclusive
Ends: 28-Dec-2026
111.00
Save 16.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
94.65
Special offer
Online exclusive
Ends: 28-Dec-2026
111.00
Save 16.35
Each
Chemical Identifiers
CAS1305-62-0
IUPAC Namecalcium dihydroxide
Molecular FormulaCaH2O2
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Arsenic (As)=<3 ppm
Mg and alkali salts (SO4)=<2.5 %
Appearance (Color)Off-white
Heavy metals (as Pb)=<40 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only