Metaphosphoric acid, stabilized
Metaphosphoric acid, stabilized
Metaphosphoric acid, stabilized
Thermo Scientific Chemicals

Metaphosphoric acid, stabilized

Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max. | CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 219221000
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Chemical Identifiers
CAS37267-86-0
IUPAC Nametrihydroxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular FormulaH3O9P3
InChI KeyAZSFNUJOCKMOGB-UHFFFAOYSA-N
SMILESOP1(=O)OP(O)(=O)OP(O)(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Arsenic (As)=<5 ppm
Appearance (Form)Lumps
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<100 ppm
Appearance (Color)Colorless to white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only