Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals
Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals
Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals

数量:
25 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 219235000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7789-29-9
IUPAC Namepotassium hydrogen difluoride
Molecular FormulaF2HK
InChI KeyVBKNTGMWIPUCRF-UHFFFAOYSA-M
SMILES[H+].[F-].[F-].[K+]
さらに表示
Appearance (Form)Crystalline powder or crystals
Titration with NaOH99.0 to 101.0 %
Water=<0.3 % (KF)
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<50 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only