Sodium sulfate, 99+%, for analysis, crystalline, anhydrous
Sodium sulfate, 99+%, for analysis, crystalline, anhydrous
Sodium sulfate, 99+%, for analysis, crystalline, anhydrous
Thermo Scientific Chemicals

Sodium sulfate, 99+%, for analysis, crystalline, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 219260025
Price (USD)
260.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
260.00
Each
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Magnesium (Mg)=<50 ppm
Phosphate (PO4)=<20 ppm
Potassium (K)=<100 ppm
Calcium (Ca)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only