N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylethylamine, 99%

CAS: 598-56-1 | C4H11N | 73.13 g/mol
数量:
250 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 219292500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS598-56-1
IUPAC Nameethyldimethylamine
Molecular FormulaC4H11N
InChI KeyDAZXVJBJRMWXJP-UHFFFAOYSA-N
SMILESCCN(C)C
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.3715 to 1.3735 (20°C, 589 nm)
Appearance (Form)Liquid
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only