Search
View additional product information for EZ-Link™ Maleimide-PEG Solid-Phase Biotinylation Kit - Mini-Spin Columns - FAQs (21930)
1 product FAQs found
The standard biotinylation kits contain components for labeling 1-10 mg of protein, and the Micro Biotinylation Kits are structured for labeling 50-200 µg of protein in 200-700 mL. The Micro kits contain the biotinylation reagents in small, individual packages and do not include components to perform the HABA assay.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.