Hydroquinone, 99.5%
Hydroquinone, 99.5%
Hydroquinone, 99.5%
Thermo Scientific Chemicals

Hydroquinone, 99.5%

CAS: 123-31-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 219930010
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Chemical Identifiers
CAS123-31-9
IUPAC Namebenzene-1,4-diol
Molecular FormulaC6H6O2
InChI KeyQIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or needles
Appearance (Color)White to off-white
Melting point171°C to 174°C
Sulfated ash=<0.05 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only