Propionic acid, 99+%, extra pure
Propionic acid, 99+%, extra pure
Propionic acid, 99+%, extra pure
Thermo Scientific Chemicals

Propionic acid, 99+%, extra pure

CAS: 79-09-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 220130025
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
166.00
Save 24.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
166.00
Save 24.35
Each
Chemical Identifiers
CAS79-09-4
IUPAC Namepropanoic acid
Molecular FormulaC3H6O2
InChI KeyXBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILESCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.1 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.3850 to 1.3870 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only