Ethyl acetoacetate, 99+%, extra pure
Ethyl acetoacetate, 99+%, extra pure
Ethyl acetoacetate, 99+%, extra pure
Thermo Scientific Chemicals

Ethyl acetoacetate, 99+%, extra pure

CAS: 141-97-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 220401000
Price (USD)
41.10
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.10
Each
Chemical Identifiers
CAS141-97-9
IUPAC Nameethyl 3-oxobutanoate
Molecular FormulaC6H10O3
InChI KeyXYIBRDXRRQCHLP-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only