Tetraethylammonium bromide, 99+%
Tetraethylammonium bromide, 99+%
Tetraethylammonium bromide, 99+%
Thermo Scientific Chemicals

Tetraethylammonium bromide, 99+%

CAS: 71-91-0 | C8H20BrN | 210.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 220712500
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS71-91-0
IUPAC Nametetraethylazanium bromide
Molecular FormulaC8H20BrN
InChI KeyHWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES[Br-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration Argentometric>=99.0 %
Appearance (Color)White to cream
Loss on drying=<1.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only