Gallic acid monohydrate
Gallic acid monohydrate
Gallic acid monohydrate
Thermo Scientific Chemicals

Gallic acid monohydrate

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 220760025
Price (USD)
692.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
692.00
Each
Chemical Identifiers
CAS5995-86-8
IUPAC Name3,4,5-trihydroxybenzoate
Molecular FormulaC7H5O5
InChI KeyLNTHITQWFMADLM-UHFFFAOYSA-M
SMILESOC1=CC(=CC(O)=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 % (120°C)
Titration with NaOH>=99 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only