1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
Thermo Scientific Chemicals

1,2-Propanediol, 99+%, for analysis

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 220870010
Price (USD)
163.65
Online exclusive
182.00
Save 18.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
163.65
Online exclusive
182.00
Save 18.35
Each
Chemical Identifiers
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (Coulometric)
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Chloride (Cl)=<50 ppm
Lead (Pb)=<0.1 ppm
Copper (Cu)=<0.1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only