Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 220982500
Price (USD)
-
Request bulk or custom format
Chemical Identifiers
CAS54-21-7
IUPAC Namesodium 2-hydroxybenzoate
Molecular FormulaC7H5NaO3
InChI KeyABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES[Na+].OC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Titration with HClO499.0 to 101.0 % (on dried substance)
Infrared spectrumConforms
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Powder or crystals or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only