N-Phenyl-p-phenylenediamine, 98%
N-Phenyl-p-phenylenediamine, 98%
N-Phenyl-p-phenylenediamine, 98%
Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%

CAS: 101-54-2 | C12H12N2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 221000010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS101-54-2
IUPAC NameN1-phenylbenzene-1,4-diamine
Molecular FormulaC12H12N2
InChI KeyATGUVEKSASEFFO-UHFFFAOYSA-N
SMILESNC1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown to purple
Appearance (Form)Powder or crystals or flakes or mass and/or chunks
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only