4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals

CAS: 2493-02-9 | Molecular Formula: C7H4BrNO | Molecular Weight: 198.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
22113010010 gGlass bottle
Catalog number 221130100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2493-02-9
IUPAC Name1-bromo-4-isocyanatobenzene
Molecular FormulaC7H4BrNO
InChI KeyCZQIJQFTRGDODI-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline solid
Melting point39°C to 42°C
Infrared spectrumConforms
GC>=98.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only