1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%
Thermo Scientific Chemicals

1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%

CAS: 74257-00-4 | C11H12N4O4S | 296.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221310050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS74257-00-4
IUPAC Name3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Molecular FormulaC11H12N4O4S
InChI KeySFYDWLYPIXHPML-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=97.5 %
Melting point131°C to 139°C
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only