Potassium perchlorate, ACS reagent
Potassium perchlorate, ACS reagent
Potassium perchlorate, ACS reagent
Thermo Scientific Chemicals

Potassium perchlorate, ACS reagent

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221371000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7778-74-7
IUPAC Namepotassium perchlorate
Molecular FormulaClKO4
InChI KeyYLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Insoluble matter=<0.005 %
Sulfate (SO4)=<0.001 %
Chloride (Cl)=<0.003 %
Calcium (Ca)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only