Product Image
Thermo Scientific™
Pressure Cells for HAAKE™ Rheometers
Our comprehensive pressure cell portfolio allows for tests at pressures up to 600 bar and temperatures up to 300 °C.
Have Questions?
Catalog NumberDescription
222-2284Pressure Cell D60/200 (up to 60 bar and 200 °C) made of titanium
Catalog number 222-2284
Price (MXN)
-
Description:
Pressure Cell D60/200 (up to 60 bar and 200 °C) made of titanium
Conduct rheological tests at temperatures of up to 300 °C and pressures up to 600 bars by adding pressure cells and temperature control modules to your rheometer. The Thermo Scientific™ HAAKE™ MARS™ and Thermo Scientific™ HAAKE™ Viscotester iQ™ Rheometers can be equipped with different modules and cells for testing under various conditions.
Enable your laboratory to perform rheological tests at elevated pressures by taking advantage of our portfolio of pressure cells for HAAKE MARS and HAAKE Viscotester iQ rheometer platforms. Features include:
  • Powerful magnetic coupling between motor drive shaft and measuring geometry for high torque measurements
  • Measuring geometries with low friction sapphire-bearing for performing low torque measurements
  • Pressure cells and measuring geometries made of titanium or Hastelloy™ for high chemical resistance
  • Large sample volume for representative test results for inhomogeneous materials
  • Broad variety of cylinder and vane rotor geometries
Specifications
TypePressure Cell
DescriptionPressure Cell D60/200 (up to 60 bar and 200 °C) made of titanium
Unit SizeEach