1,2-Dichlorobenzene, 99+%, for spectroscopy
1,2-Dichlorobenzene, 99+%, for spectroscopy
1,2-Dichlorobenzene, 99+%, for spectroscopy
Thermo Scientific Chemicals

1,2-Dichlorobenzene, 99+%, for spectroscopy

CAS: 95-50-1 | C6H4Cl2 | 147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 222050025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 %
UV1 cm cell vs water
UVat 296 nm A: =<1
UVat 300 nm A: =<0.3
UVat 336 nm A: =<0.05
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
RUO – Research Use Only