Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals
Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals
Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 222192500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-48-3
IUPAC Nameethyl 2-iodoacetate
Molecular FormulaC4H7IO2
InChI KeyMFFXVVHUKRKXCI-UHFFFAOYSA-N
SMILESCCOC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to orange
Refractive index1.5025 to 1.5055 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only