Chromium potassium sulfate dodecahydrate, 98+%
Chromium potassium sulfate dodecahydrate, 98+%
Chromium potassium sulfate dodecahydrate, 98+%
Thermo Scientific Chemicals

Chromium potassium sulfate dodecahydrate, 98+%

CAS: 7788-99-0 | CrKO8S2·12H2O | 499.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 222520025
Price (EUR)
276,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
276,00
Each
Chemical Identifiers
CAS7788-99-0
IUPAC Namechromium(3+) potassium dodecahydrate disulfate
Molecular FormulaCrH24KO20S2
InChI KeyZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Violet to dark purple
Appearance (Form)Crystalline powder or crystals
Titration Iodometric98.0 to 102.0 %
Chloride (Cl)=<20 ppm
Ammonium (NH4)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only