Manganese(IV) Oxide, 99%, -10 mesh
Manganese(IV) Oxide, 99%, -10 mesh
Manganese(IV) Oxide, 99%, -10 mesh
Thermo Scientific Chemicals

Manganese(IV) Oxide, 99%, -10 mesh

CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 222580025
Price (USD)
515.65
Special offer
Online exclusive
Ends: 28-Dec-2026
607.00
Save 91.35
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
515.65
Special offer
Online exclusive
Ends: 28-Dec-2026
607.00
Save 91.35
Each
Chemical Identifiers
CAS1313-13-9
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
Appearance (Form)Powder
Titration with Na2S2O3>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only