Search
View additional product information for Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle - FAQs (2226-0020, 2226-0050PK, 2226-0050)
3 product FAQs found
These bottles can hold full vacuum for up to 24 hrs at 20 degrees C.
Nalgene polypropylene carboys are safe to lift by the handles when full, as they are capable of supporting at least double their intended weight on a single handle.
To test the strength of the carboy handles, carboys are suspended through a single handle on the carboy body. 10 L, 20 L, and 50 L carboys are tested by filling the container with steel ballast pellets to a weight in excess of double the weight of the carboy filled to nominal capacity with water.
Unfortunately, none of our containers were designed or tested for applications involving positive pressure. Some of our customers do choose to carry out their own performance validations, but this must be done at the customers own risk. We do offer selected bottles and carboys (2126 and 2226 series) that are vacuum rated for use at room temperature (to full rough vacuum, about 28-29 in. Hg).