Ammonium sodium phosphate tetrahydrate, 99%, for analysis
Ammonium sodium phosphate tetrahydrate, 99%, for analysis
Ammonium sodium phosphate tetrahydrate, 99%, for analysis
Thermo Scientific Chemicals

Ammonium sodium phosphate tetrahydrate, 99%, for analysis

CAS: 7783-13-3 | H5NNaO4P·4H2O | 209.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 222642500
Price (USD)
33.65
Online exclusive
37.00
Save 3.35
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
33.65
Online exclusive
37.00
Save 3.35
Each
Chemical Identifiers
CAS7783-13-3
IUPAC Nameammonium sodium tetrahydrate hydrogen phosphate
Molecular FormulaH13NNaO8P
InChI KeyIQTQISLCLWJRPM-UHFFFAOYSA-M
SMILES[NH4+].O.O.O.O.[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<10 ppm
Appearance (Form)Crystalline powder or crystals
Nitrate (NO3)=<20 ppm
Arsenic (As)=<1 ppm
Appearance (Color)Colorless to white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only