Potassium nitrite, 97%, for analysis
Potassium nitrite, 97%, for analysis
Potassium nitrite, 97%, for analysis
Thermo Scientific Chemicals

Potassium nitrite, 97%, for analysis

CAS: 7758-09-0 | KNO2 | 85.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 222702500
Price (USD)
80.30
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.30
Each
Chemical Identifiers
CAS7758-09-0
IUPAC Namepotassium nitrite
Molecular FormulaKNO2
InChI KeyBXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES[K+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO4>=96.0 %
Iron (Fe)=<10 ppm
Chloride (Cl)=<100 ppm
Appearance (Form)Crystals
Arsenic (As)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only