Magnesium chloride, pure
Magnesium chloride, pure
Magnesium chloride, pure
Thermo Scientific Chemicals

Magnesium chloride, pure

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 223210010
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.80
Save 10.15
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.80
Save 10.15
Each
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain grey to black particles
Appearance (Color)White
Water=<3 % (K.F.)
Appearance (Form)Powder
Titration Complexometric>=98.0 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only