Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals

数量:
25 mL
500 mL
2.5 L
パッケージ:
Plastic bottle
製品番号(カタログ番号) 223335000
価格(JPY)
-
数量:
500 mL
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7664-39-3
IUPAC Namehydrogen fluoride
Molecular FormulaFH
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
さらに表示
Titration with NaOH48 to 51 %
Chloride (Cl)=<5 ppm
Phosphates=<1 ppm (PO4)
Copper (Cu)=<0.1 ppm
Lead (Pb)=<0.05 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only