Methyl 2-butynoate, 97%
Methyl 2-butynoate, 97%
Methyl 2-butynoate, 97%
Thermo Scientific Chemicals

Methyl 2-butynoate, 97%

CAS: 23326-27-4 | C5H6O2 | 98.1 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 223490010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS23326-27-4
IUPAC Namemethyl but-2-ynoate
Molecular FormulaC5H6O2
InChI KeyUJQCANQILFWSDJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC
さらに表示
Appearance (Form)Liquid
Refractive index1.4360 to 1.4380 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only