hydrogen tetrachloroaurate(III) hydrate
hydrogen tetrachloroaurate(III) hydrate
hydrogen tetrachloroaurate(III) hydrate
Acros Organics

hydrogen tetrachloroaurate(III) hydrate

CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 223620050
HSN 28433000
Price (INR)
130,240.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
130,240.00
Each
Chemical Identifiers
CAS27988-77-8
IUPAC Namehydrogen tetrachlorogoldtris(ylium)
Molecular FormulaAuCl4H
InChI KeyRFWCEGCWRSNAGT-UHFFFAOYSA-K
SMILES[H+].Cl[Au+3](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.1 % (in diethyl ether)
Appearance (Color)Yellow to orange
Gold (Au)>=49 %
Alkalies+oth.metals (SO4)=<0.2 %
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only