Tin(IV) chloride pentahydrate, 98+%, extra pure
Tin(IV) chloride pentahydrate, 98+%, extra pure
Tin(IV) chloride pentahydrate, 98+%, extra pure
Thermo Scientific Chemicals

Tin(IV) chloride pentahydrate, 98+%, extra pure

CAS: 10026-06-9 | Cl4H10O5Sn | 350.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 223695000
Price (USD)
179.65
Online exclusive
199.00
Save 19.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
179.65
Online exclusive
199.00
Save 19.35
Each
Chemical Identifiers
CAS10026-06-9
IUPAC Nametin(4+) pentahydrate tetrachloride
Molecular FormulaCl4H10O5Sn
InChI KeyKHMOASUYFVRATF-UHFFFAOYSA-J
SMILESO.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or lumps
Iron (Fe)=<50 ppm
Assay>=98.0 %
Appearance (Color)White to off-white
Lead (Pb)=<50 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only