1-Bromoundecane, 98%
1-Bromoundecane, 98%
1-Bromoundecane, 98%
Thermo Scientific Chemicals

1-Bromoundecane, 98%

CAS: 693-67-4 | C11H23Br | 235.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 224010250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS693-67-4
IUPAC Name1-bromoundecane
Molecular FormulaC11H23Br
InChI KeyIKPSIIAXIDAQLG-UHFFFAOYSA-N
SMILESCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4560 to 1.4580 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only